Anti-NOG Mouse Monoclonal Antibody [clone: OTI1C1]

Supplier: OriGene
TA500116
ORIGTA500116EA 577 GBP
ORIGTA500116
Anti-NOG Mouse Monoclonal Antibody [clone: OTI1C1]
Antibodies
Recommended Dilutions: Western Blot: 1:40000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NOG (noggin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR