Anti-PITPNM3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308307EA 614 GBP
ORIGTA308307
Anti-PITPNM3 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: PITPNM3 (PITPNM family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR