Anti-ACTC1 Rabbit Monoclonal Antibody [clone: EP184E]

Supplier: OriGene
ORIGTA303378EA 790 GBP
ORIGTA303378
Anti-ACTC1 Rabbit Monoclonal Antibody [clone: EP184E]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100-500; Flow Cytometry: 1:20; Immunoprecipitation: 1:20

Type: Primary
Antigen: ACTC1 (actin, alpha, cardiac muscle 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR