Anti-HNF1B Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302622EA 513 GBP
ORIGTA302622
Anti-HNF1B Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-0.3µg/ml.An

Type: Primary
Antigen: HNF1B (HNF1 homeobox B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR