Anti-IL-23/IL-12p40 Rat Monoclonal Antibody [clone: C17.8]

Supplier: Tonbo Biosciences
40-7123-U500 40-7123-U100
40-7123-U500EA 178 GBP
40-7123-U500 40-7123-U100
Anti-IL-23/IL-12p40 Rat Monoclonal Antibody [clone: C17.8]
Antibodies
The C17.8 antibody is specific for the 40 kDa (p40) protein subunit shared by the cytokines IL-12 and IL-23. To form IL-12, p40 assembles with a separate 35 kDa protein known as p35, resulting in a 70 kDa functional cytokine. IL-12 is secreted by activated monocytes, macrophages, and dendritic cells, and has been shown to target naïve, resting CD4+ T cells to promote their proliferation and secretion of cytokines. IL-23 contains the p40 subunit in combination with a 19 kDa protein chain, p19; its primary source being activated dendritic cells and other antigen-presenting cells. IL-23 appears to target different cell types than IL-12, acting on memory CD4+ T cells to induce a strong proliferative response and contributing to the generation and expansion of Th17 cells. Like the cytokines themselves, the receptors for IL-12 and IL-23 share one subunit, as well as containing distinct cytokine-specific subunits.

As the C17.8 antibody binds to a shared subunit of both IL-12 and IL-23, it may be used as a marker for either IL-12 or IL-23 expression in dendritic cells, monocytes and macrophages, and is widely used for neutralization of activity associated with either cytokine. Please choose the appropriate format for each application.

Type: Primary
Antigen: IL-23/IL-12p40 (IL-23/IL-12p40)
Clonality: monoclonal
Clone: C17.8
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR