Anti-EGR2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
P100880_P050
AVIVP100880_P050EA 339 GBP
AVIVP100880_P050
Anti-EGR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EGR2 (early growth response 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR