Anti-IGLON5 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP70564_P050
AVIVARP70564_P050EA 339 GBP
AVIVARP70564_P050
Anti-IGLON5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IGLON5 (IgLON family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR