Anti-UBTF Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP39067_P050EA 339 GBP
AVIVARP39067_P050
Anti-UBTF Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UBTF (upstream binding transcription factor, RNA polymerase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR