Anti-ACTN2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP37422_T100
AVIVARP37422_T100EA 295 GBP
AVIVARP37422_T100
Anti-ACTN2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACTN2 (actinin, alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR