Anti-DNAJB12 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP37278_P050
AVIVARP37278_P050EA 339 GBP
AVIVARP37278_P050
Anti-DNAJB12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNAJB12 (DnaJ (Hsp40) homolog, subfamily B, member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR