Anti-KEAP1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP34728_T100EA 295 GBP
AVIVARP34728_T100
Anti-KEAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KEAP1 (kelch-like ECH-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR