Anti-PCK1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP32801_P050 ARP32801_T100
AVIVARP32801_P050EA 353 GBP
AVIVARP32801_P050 AVIVARP32801_T100
Anti-PCK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PCK1 (phosphoenolpyruvate carboxykinase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR