Adapters, reduction, for conical ground joints
Supplier: SMITH SCIENTIFIC
8SRA01 8SRA12 8SRA13 8SRA14 8SRA23 8SRA24 8SRA25 8SRA26 8SRA27 8SRA34 8SRA35 8SRA36 8SRA37 8SRA38 8SRA45 8SRA47 8SRA56 8SRA58
SMIT8SRA01EA 96.5 GBP
SMIT8SRA01 SMIT8SRA12 SMIT8SRA13 SMIT8SRA14 SMIT8SRA23 SMIT8SRA24 SMIT8SRA25 SMIT8SRA26 SMIT8SRA27 SMIT8SRA34 SMIT8SRA35 SMIT8SRA36 SMIT8SRA37 SMIT8SRA38 SMIT8SRA45 SMIT8SRA47 SMIT8SRA56 SMIT8SRA58
Adapters, reduction, for conical ground joints
Adapters Adapters, Conical Ground Joints
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.