Liebig condenser with standard ground joints
Supplier: SMITH SCIENTIFIC LTD
8SCL1/11/SC 8SCL1/22/200/ 8SCL1/33/SC 8SCL2/22/SC 8SCL2/33/SC
SMIT8SCL1/11/SCEA 113 GBP
SMIT8SCL1/11/SC SMIT8SCL1/22/200/ SMIT8SCL1/33/SC SMIT8SCL2/22/SC SMIT8SCL2/33/SC
Liebig condenser with standard ground joints
Condensers
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With screw thread connectors.