Condensers, Graham
Supplier: SMITH SCIENTIFIC LTD
8SCC20/B19/SC 8SCC25/B24/SC 8SCC30/B19/SC 8SCC30/B24/SC
SMIT8SCC20/B19/SCEA 445 GBP
SMIT8SCC20/B19/SC SMIT8SCC25/B24/SC SMIT8SCC30/B19/SC SMIT8SCC30/B24/SC
Condensers, Graham
Condensers
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graham coil condensers with screw thread tubing connectors.