Condensers, Allihn
Supplier: SMITH SCIENTIFIC
8SCAL20/B34/S 8SCAL20/B40/S 8SCAL20/B50/S
SMIT8SCAL20/B34/SEA 0 GBP
SMIT8SCAL20/B34/S SMIT8SCAL20/B40/S SMIT8SCAL20/B50/S
Condensers, Allihn
Condensers
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Allihn condensers for condensing vapours, with cone, water connection with screw thread connectors.