Air condensers
Supplier: SMITH SCIENTIFIC LTD
8SCA12/11 8SCA12/12 8SCA12/22 8SCA12/23 8SCA12/43 8SCA12/44
SMIT8SCA12/11EA 49.7 GBP
SMIT8SCA12/11 SMIT8SCA12/12 SMIT8SCA12/22 SMIT8SCA12/23 SMIT8SCA12/43 SMIT8SCA12/44
Air condensers
Condensers
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With cone and socket.