Methyl-6-fluoronicotinate 95%

Supplier: Thermo Scientific
Danger

Synonyms: 6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate

H61302.MD H61302.03
H61302.MDEA 54.1 GBP
H61302.MD H61302.03
Methyl-6-fluoronicotinate 95%
Methyl 6-fluoronicotinate
Formula: C₇H₆FNO₂
MW: 155,13 g/mol
Melting Pt: 48…52 °C
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD03095098
CAS Number: 1427-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR