2,3-Dichloroisonicotinic acid ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,3-Dichloropyridine-4-carboxylic acid
H63733.03
H63733.06
H63733.03EA
0
GBP
H63733.03
H63733.06
2,3-Dichloroisonicotinic acid ≥97%
2,3-Dichloroisonicotinic acid
Purity: 97%
CAS No.: 184416-84-0
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Physical Form: Solid
Melting Point: 203-207°
MDL No.: MFCD01861977
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 203…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861977 CAS Number: 184416-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...