4-Amino-2-fluorophenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

H59916.MD H59916.03
H59916.MDEA 69.8 GBP
H59916.MD H59916.03
4-Amino-2-fluorophenylboronic acid pinacol ester 96%
4-Amino-2-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₂
MW: 237,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09951877
CAS Number: 819057-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR