1-Methyl-3-octylimidazolium hexafluorophosphate 95%

Supplier: Thermo Scientific
Warning

H59990.14
H59990.14EA 259 GBP
H59990.14
1-Methyl-3-octylimidazolium hexafluorophosphate 95%
1-Methyl-3-octylimidazolium hexafluorophosphate
Formula: C₁₂H₂₃F₆N₂P
MW: 340,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798203
CAS Number: 304680-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR