5-Methyl-1H-tetrazole ≥97%

Supplier: Thermo Scientific
Warning

H60248.06 H60248.14
H60248.06EA 50.7 GBP
H60248.06 H60248.14
5-Methyl-1H-tetrazole ≥97%
5-Methyl-1H-tetrazole
Formula: C₂H₄N₄
MW: 84,08 g/mol
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00185824
CAS Number: 4076-36-2
EINECS: 218-746-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR