(2,6-Dichlorophenylthio)acetic acid ≥99%

Supplier: Thermo Scientific
Warning

H50282.03 H50282.06
H50282.03EA 88.9 GBP
H50282.03 H50282.06
(2,6-Dichlorophenylthio)acetic acid ≥99%
(2,6-Dichlorophenylthio)acetic acid

Purity: 99% CAS No.: 21248-45-3 Molecular Formula: C8H6Cl2O2S Formula Weight: 237.11 MDL No.: MFCD00052108

Formula: C₈H₆Cl₂O₂S
MW: 237,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052108
CAS Number: 21248-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR