2-Cyclopentylethanol 98

Supplier: Thermo Fisher Scientific
Warning

B22344.03 B22344.06
B22344.03EA 64.5 GBP
B22344.03 B22344.06
2-Cyclopentylethanol 98
2-Cyclopentylethanol

Purity: 98+% CAS No.: 766-00-7 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 70°(158°F) Refractive Index: 1.4585 Boiling Point: 182° Beilstein Reference: 2069860 MDL No.: MFCD00019301

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 182 °C
Density: 0.920
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019301
CAS Number: 766-00-7
EINECS: 212-156-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR