Cyclobutyl methyl ketone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Cyclobutylethan-1-one

L03521.03 L03521.06
L03521.03EA 34.9 GBP
L03521.03 L03521.06
Cyclobutyl methyl ketone ≥97%
Cyclobutyl methyl ketone

Purity: 97% CAS No.: 3019-25-8 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 28°(82°F) Refractive Index: 1.4320 Boiling Point: 134-135° Beilstein Reference: 1924022 UN No.: 1224 MDL No.: MFCD00037148

Formula: C₆H₁₀O
MW: 98,15 g/mol
Boiling Pt: 134…135 °C
Density: 0,900
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037148
CAS Number: 3019-25-8
EINECS: 221-163-6
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR