N,N-Diisopropylaniline ≥97%

Supplier: Thermo Scientific
Warning

H55471.03 H55471.06
H55471.03EA 51.3 GBP
H55471.03 H55471.06
N,N-Diisopropylaniline ≥97%
N,N-Diisopropylaniline

Purity: 97% CAS No.: 4107-98-6 Molecular Formula: C12H19N Formula Weight: 177.29 Flash Point: 84°(183°F) Physical Form: Liquid Density: 0.91 Refractive Index: 1.5190 Sensitivity: Light Sensitive Appearance: Colourless to pale yellow Boiling Point: 95-96°/11mm MDL No.: MFCD00048277

Formula: C₁₂H₁₉N
MW: 177,29 g/mol
Boiling Pt: 95…96 °C (11 mmHg)
Density: 0,91
Flash Pt: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048277
CAS Number: 4107-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR