Diheptyltin dichloride

Supplier: LGC Standards PROMOCHEM

PROODEMS-03442-0.3EA 237 GBP
PROODEMS-03442-0.3
Diheptyltin dichloride
Diheptyltin dichloride
Formula: C₁₄H₃₀Cl₂Sn
CAS Number: 74340-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR