5-Acetyl-2-methylpyridine ≥97%

Supplier: Thermo Scientific
Warning

H27014.03 H27014.MD
H27014.03EA 0 GBP
H27014.03 H27014.MD
5-Acetyl-2-methylpyridine ≥97%
5-Acetyl-2-methylpyridine

Purity: 97% CAS No.: 36357-38-7 Molecular Formula: C8H9NO Formula Weight: 135.17 MDL No.: MFCD00128113

Formula: C₈H₉NO
MW: 135,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128113
CAS Number: 36357-38-7
EINECS: 252-995-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR