N,N-Dimethyl-4-(2-pyridylazo)aniline ≥98%
Supplier: Thermo Scientific
|
Warning
|
B23789.03
B23789.06
B23789.03EA
0
GBP
B23789.03
B23789.06
N,N-Dimethyl-4-(2-pyridylazo)aniline ≥98%
N,N-Dimethyl-4-(2-pyridylazo)aniline
Purity: 98%
CAS No.: 13103-75-8
Molecular Formula: C13H14N4
Formula Weight: 226.28
Melting Point: 108-111°
Beilstein Reference: 185623
Formula:
C₁₃H₁₄N₄ MW: 226,28 g/mol Melting Pt: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010435 CAS Number: 13103-75-8 EINECS: 236-026-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...