4-Amino-8-(trifluoromethyl)quinoline 97%

Supplier: Thermo Scientific
Warning

Synonyms: 8-(Trifluoromethyl)quinolin-4-amine

H50193.03 H50193.MD
H50193.03EA 162 GBP
H50193.03 H50193.MD
4-Amino-8-(trifluoromethyl)quinoline 97%
4-Amino-8-(trifluoromethyl)quinoline

Purity: 97% CAS No.: 243977-15-3 Molecular Formula: C10H7F3N2 Formula Weight: 212.18 MDL No.: MFCD00269346

Formula: C₁₀H₇F₃N₂
MW: 212,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269346
CAS Number: 243977-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR