(Furfurylthio)acetone 96%

Supplier: Thermo Scientific
Warning

Synonyms: 1-((Furan-2-ylmethyl)thio)propan-2-one

B22724.06 B22724.14
B22724.06EA 37.6 GBP
B22724.06 B22724.14
(Furfurylthio)acetone 96%
(Furfurylthio)acetone

Purity: 96% CAS No.: 58066-86-7 Molecular Formula: C8H10O2S Formula Weight: 170.23 MDL No.: MFCD02683098

Formula: C₈H₁₀O₂S
MW: 170,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683098
CAS Number: 58066-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR