4-(Trifluoromethyl)piperidine hydrochloride ≥97%
Supplier: Thermo Scientific
|
Warning
|
L19441.03
L19441.MD
L19441.03EA
132
GBP
L19441.03
L19441.MD
4-(Trifluoromethyl)piperidine hydrochloride ≥97%
4-(Trifluoromethyl)piperidine hydrochloride
Purity: 97%
CAS No.: 155849-49-3
Molecular Formula: C6H10F3N.HCl
Formula Weight: 189.61
Melting Point: ≈160° dec.
Beilstein Reference: 1341222
Formula:
C₆H₁₁ClF₃N MW: 189,61 g/mol Melting Pt: ∼160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04971993 CAS Number: 155849-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...