2-(Tributylstannyl)furan ≥97%

Supplier: Thermo Scientific
Danger

H55259.AA H55259.AC
H55259.AAEA 30.6 GBP
H55259.AA H55259.AC
2-(Tributylstannyl)furan ≥97%
2-(Tributylstannyl)furan
Formula: C₁₆H₃₀OSn
MW: 357,10 g/mol
Boiling Pt: 90 °C (0,1 mmHg)
Density: 1,134
Storage Temperature: Ambient
MDL Number: MFCD00192512
CAS Number: 118486-94-5
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR