2,2'-Biquinoline 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
106300010
106300050
ACRO106300010EA
65.2
GBP
ACRO106300010
ACRO106300050
2,2'-Biquinoline 98%
2,2'-Biquinoline
Formula:
C₁₈H₁₂N₂ MW: 256.31 g/mol Melting Pt: 193…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006740 CAS Number: 119-91-5 EINECS: 204-357-5 |
Specification Test Results
Appearance | White to cream to yellow to light brown Powder or crystals or flakes and/or chunks |
Infrared spectrum | Conforms |
Melting point | 192 to 198 °C |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...