Tantalum(V) tetraethoxide 2,4-pentanedionate ≥99.99% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

39630.04 39630.09 39630.18
39630.04EA 83.2 GBP
39630.04 39630.09 39630.18
Tantalum(V) tetraethoxide 2,4-pentanedionate ≥99.99% (metals basis)

Purity: 99.99%, CAS No.: 20219-33-4 Molecular Formula: C13H27O6Ta Formula Weight: 460.30 Flash Point: 65°(149°F) Physical Form: Solid Density: 1.5 Sensitivity: Moisture Sensitive Melting Point: 45° Boiling Point: 95°/0.5mm MDL No.: MFCD00145349

Formula: C₁₃H₂₇O₆Ta
Boiling Pt: 95 °C (0,5 mmHg)
Melting Pt: 45 °C
Density: 1,5
Flash Pt: >65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00145349
CAS Number: 20219-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR