N-Fmoc-D-valine ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Fmoc-D-Val-OH, D-Valine, N-FMOC protected

H59570.03 H59570.06
H59570.03EA 0 GBP
H59570.03 H59570.06
N-Fmoc-D-valine ≥98%
N-Fmoc-D-valine
Formula: C₂₀H₂₁NO₄
Melting Pt: 143…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062953
CAS Number: 84624-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR