1-Allylimidazole ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L11053.09
L11053.18
L11053.09EA
23.6
GBP
L11053.09
L11053.18
1-Allylimidazole ≥97%
1-Allylimidazole
Purity: 99%
CAS No.: 31410-01-2
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 101°(213°F)
Refractive Index: 1.5050
Boiling Point: 212-213°
Beilstein Reference: 605953
UN No.: 3267
MDL No.: MFCD00037894
Formula:
C₆H₈N₂ Boiling Pt: 212…213 °C Density: 1,003 Flash Pt: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037894 CAS Number: 31410-01-2 EINECS: 250-618-1 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...