4,6-Dichloroquinoline ≥97%

Supplier: Thermo Scientific
Warning

H59307.MD H59307.03
H59307.MDEA 48.2 GBP
H59307.MD H59307.03
4,6-Dichloroquinoline ≥97%
4,6-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198,05 g/mol
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00156141
CAS Number: 4203-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR