α,2-Dibromo-4-fluorotoluene 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromo-4-fluorobenzylbromide, 2-Bromo-4-fluorobenzyl bromide

H62157.03 H62157.06
H62157.03EA 0 GBP
H62157.03 H62157.06
α,2-Dibromo-4-fluorotoluene 96%
α,2-Dibromo-4-fluorotoluene

Purity: 96% CAS No.: 61150-57-0 Molecular Formula: C7H5Br2F Formula Weight: 267.92 Physical Form: Solid UN No.: 3261 MDL No.: MFCD03840524

Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840524
CAS Number: 61150-57-0
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR