4'-Bromo-3'-fluoroacetophenone 96%

Supplier: Thermo Fisher Scientific

Synonyms: 4-Bromo-3-fluoro--acetophenone

H62040.MD H62040.06
H62040.MDEA 0 GBP
H62040.MD H62040.06
4'-Bromo-3'-fluoroacetophenone 96%
4'-Bromo-3'-fluoroacetophenone

Purity: 96% CAS No.: 304445-49-6 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466241
CAS Number: 304445-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR