4-Methyl-4-hepten-3-ol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B20658.03
B20658.06
B20658.14
B20658.03EA
0
GBP
B20658.03
B20658.06
B20658.14
4-Methyl-4-hepten-3-ol ≥97%
4-Methyl-4-hepten-3-ol
Purity: 97%
CAS No.: 81280-12-8
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 59°(138°F)
MDL No.: MFCD00046670
Formula:
C₈H₁₆O Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046670 CAS Number: 81280-12-8 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...