6-Nitrophthalide ≥97%

Supplier: Thermo Fisher Scientific
Warning

B20375.06 B20375.14
B20375.06EA 56.4 GBP
B20375.06 B20375.14
6-Nitrophthalide ≥97%
6-Nitrophthalide

Purity: 97% CAS No.: 610-93-5 Molecular Formula: C8H5NO4 Formula Weight: 179.13 Melting Point: 140-146° MDL No.: MFCD00033529

Formula: C₈H₅NO₄
Melting Pt: 140…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00033529
CAS Number: 610-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR