4-(4-Methoxyphenoxy)piperidine ≥99%

Supplier: Thermo Fisher Scientific
Warning

H50966.03 H50966.MD
H50966.03EA 0 GBP
H50966.03 H50966.MD
4-(4-Methoxyphenoxy)piperidine ≥99%
4-(4-Methoxyphenoxy)piperidine

Purity: 99% CAS No.: 162402-33-7 Molecular Formula: C12H17NO2 Formula Weight: 207.27 MDL No.: MFCD08059670

Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059670
CAS Number: 162402-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR