2-Methoxy-4-(tributylstannyl)thiazole 96%

Supplier: Thermo Scientific
Danger

H51493.MD H51493.03
H51493.MDEA 71.9 GBP
H51493.MD H51493.03
2-Methoxy-4-(tributylstannyl)thiazole 96%
2-Methoxy-4-(tributylstannyl)thiazole

Purity: 96% CAS No.: 927391-09-1 Molecular Formula: C16H31NOSSn Formula Weight: 404.20 Storage Temperature: 2°C to 8°C

Formula: C₁₆H₃₁NOSSn
MW: 404,20 g/mol
MDL Number: MFCD07787372
CAS Number: 927391-09-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR