Tungsten (platinium coated), wire, Ø 0.25 mm (0.01 in)

Supplier: Thermo Fisher Scientific

13630.G1 13630.G5
13630.G1EA 171 GBP
13630.G1 13630.G5
Tungsten (platinium coated), wire, Ø 0.25 mm (0.01 in)
Tungsten (platinium coated)

Physical Form: Pt thickness 5 micron MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR