Ethyl-3-chloro-5-(trifluoromethyl)picolinate 96%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 3-chloro-5-(trifluoromethyl)picolinate, Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

H33967.03 H33967.09 H33967.06
H33967.03EA 103 GBP
H33967.03 H33967.09 H33967.06
Ethyl-3-chloro-5-(trifluoromethyl)picolinate 96%
Ethyl 3-chloro-5-(trifluoromethyl)picolinate
Formula: C₉H₇ClF₃NO₂
MW: 253,60 g/mol
Boiling Pt: 64…66 °C (0,008 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06656414
CAS Number: 128073-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR