4-Fluoro-3-methylthiobenzamide ≥97%
Supplier: Thermo Scientific
|
Danger
|
H33778.MD
H33778.03
H33778.MDEA
38.9
GBP
H33778.MD
H33778.03
4-Fluoro-3-methylthiobenzamide ≥97%
4-Fluoro-3-methylthiobenzamide
Formula:
C₈H₈FNS MW: 169,22 g/mol Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD09757567 CAS Number: 1016775-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...