5-Fluoro-2-methylthiobenzamide ≥97%

Supplier: Thermo Fisher Scientific
Danger

H33362.03 H33362.06
H33362.03EA 0 GBP
H33362.03 H33362.06
5-Fluoro-2-methylthiobenzamide ≥97%
5-Fluoro-2-methylthiobenzamide

Purity: 97% CAS No.: 1250676-94-8 Molecular Formula: C8H8FNS Formula Weight: 169.22 Melting Point: 52-56° MDL No.: MFCD09757566

Formula: C₈H₈FNS
MW: 169.22 g/mol
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD09757566
CAS Number: 1250676-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR