2-Ethoxythiobenzamide 97

Supplier: Thermo Fisher Scientific
Danger

H33353.MD H33353.03
H33353.MDEA 0 GBP
H33353.MD H33353.03
2-Ethoxythiobenzamide 97
2-Ethoxythiobenzamide

Purity: 97% CAS No.: 725702-35-2 Molecular Formula: C9H11NOS Formula Weight: 181.25 Melting Point: 113-119° MDL No.: MFCD06090837

Formula: C₉H₁₁NOS
MW: 181.25 g/mol
Melting Pt: 113…119 °C
Storage Temperature: Ambient
MDL Number: MFCD06090837
CAS Number: 725702-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR