4-(Hydroxymethyl)benzonitrile ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Cyanobenzyl alcohol
H54283.03
H54283.06
H54283.03EA
65.7
GBP
H54283.03
H54283.06
4-(Hydroxymethyl)benzonitrile ≥97%
4-(Hydroxymethyl)benzonitrile
Purity: 97%
CAS No.: 874-89-5
Molecular Formula: C8H7NO
Formula Weight: 133.15
Melting Point: 32-36°
UN No.: 3439
MDL No.: MFCD00870633
Formula:
C₈H₇NO MW: 133,15 g/mol Melting Pt: 32…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00870633 CAS Number: 874-89-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...